missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA131223
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Major component of the transverse filaments of synaptonemal complexes (SCS), formed between homologous chromosomes during meiotic prophase.
Specifications
| SCP1 | |
| Polyclonal | |
| Unconjugated | |
| SYCP1 | |
| A coiled-coil related protein specific for synapsed regions of meiotic prophase chromosomes; AW742678; cancer/testis antigen 8; CT8; DlkI; FA1; HOM-TES-14; Ly107; MGC104417; Peg9; pG2; pref-1; SCP1; SCP-1; Sycp1; Synaptonemal complex protein 1; testicular tissue protein Li 191; ZOG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20957, 25276 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q03410, Q62209 | |
| SYCP1 | |
| A synthetic peptide made to the C-terminus of the mouse SCP1 protein sequence. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction