missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110865
This item is not returnable.
View return policy
Description
Immunogen sequence: TLGGDSTFFK SFNKCTEDDF EFPFAKTNLS KNGENIDSDP ALQKVNFLPV LEQVGNSDCH YQEGLKDSDL ENSEGLSRVY SKLYKEAEKI KKWKVSTEAE LRQKESKLQE NRKIIEAQRK AIQEL Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 62%.
Major component of the transverse filaments of synaptonemal complexes (SCS), formed between homologous chromosomes during meiotic prophase.
Specifications
| SCP1 | |
| Polyclonal | |
| Unconjugated | |
| SYCP1 | |
| A coiled-coil related protein specific for synapsed regions of meiotic prophase chromosomes; AW742678; cancer/testis antigen 8; CT8; DlkI; FA1; HOM-TES-14; Ly107; MGC104417; Peg9; pG2; pref-1; SCP1; SCP-1; Sycp1; Synaptonemal complex protein 1; testicular tissue protein Li 191; ZOG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6847 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15431 | |
| SYCP1 | |
| Recombinant Protein corresonding to Human SCP1. Recombinant protein control fragment (Product #RP-92518). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction