Learn More
SCP2 Mouse anti-Human, Clone: OTI3H1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF507254
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms. The full-length nature of all transcript variants has not been determined.Specifications
| SCP2 | |
| Monoclonal | |
| Unconjugated | |
| SCP2 | |
| NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6342 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3H1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P22307 | |
| SCP2 | |
| Full length human recombit protein of human SCP2 produced in HEK293T cell | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.