missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP3/SYCP3 Antibody (6F9C5), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23487IR
This item is not returnable.
View return policy
Description
SCP3/SYCP3 Monoclonal antibody specifically detects SCP3/SYCP3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SCP3/SYCP3 | |
| Monoclonal | |
| DyLight 755 | |
| MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3 | |
| Partial recombinant human SCP3 (between residues 1-200) expressed in E. coli [Uniprot: Q8IZU3] | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 50511 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 6F9C5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction