missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP3/SYCP3 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-231H
This item is not returnable.
View return policy
Description
SCP3/SYCP3 Polyclonal specifically detects SCP3/SYCP3 in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| SCP3/SYCP3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Chicken | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| HRP | |
| PBS with No Preservative | |
| SYCP3 | |
| A synthetic peptide made to the C-terminal region of the mouse SCP3 protein. [UniProt# P70281] | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 50511 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction