Learn More
DWK Life Sciences Screw Cap with Aperture, for safety standard ground sockets
For fixing the DURAN™ core in the DURAN™ threaded sleeve.
Brand: DWK Life Sciences 286215001
15.25 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.06000kg
Description
The open screw cap reliably fixes the core of the DURAN™ security cuts in the sleeve. When the screw connection is opened, the ground joints are automatically separated from each other.
- Reliable fixation of the DURAN™ safety sections in one another (core and sleeve)
- When the screw connection is opened, the ground joints are automatically separated from each other
Specifications
10/Pk. | |
Suitable for fixing DURAN safety grinding | |
NS 14/23 |