Learn More
DWK Life Sciences Screw Cap with Aperture, for safety standard ground sockets
For fixing the DURAN™ core in the DURAN™ threaded sleeve.
Brand:  DWK Life Sciences 286215704
91.77 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.36000kg
 View more versions of this product
Product Code. 15951551
  • € 96.60 / Pack of 10
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Description
Description
The open screw cap reliably fixes the core of the DURAN™ security cuts in the sleeve. When the screw connection is opened, the ground joints are automatically separated from each other.
  • Reliable fixation of the DURAN™ safety sections in one another (core and sleeve)
  • When the screw connection is opened, the ground joints are automatically separated from each other
Specifications
Specifications
10/Pk.
Suitable for fixing DURAN safety grinding
NS 45/40