Learn More
Buerkle™ Screw Cap, Electrically Conductive, DIN50
Shop All Bürkle Products
Click to view available options
Thread Size (Metric):
30 mm
Unit Size:
Each
Description
- Made of electrically conductive HDPE
- Suitable for DIN50 thread
Specifications
Specifications
| Color | Black |
| Material | HDPE |
| Sterility | Non-sterile |
| Thread Size (Metric) | 30 mm |
| Neck Finish | 30 mm |
| Disposable | Reusable |
| Product Type | Screw Thread Cap |
| Autoclavable | Not Autoclavable |