Learn More
Buerkle™ Screw Cap, Electrically Conductive, DIN50
Product Code. 30276789 Shop All Bürkle Products
Click to view available options
Thread Size (Metric):
30 mm
Unit Size:
Each
Product Code. 30276789
Brand: Buerkle™ 03000007
Only null left
Add to basket
Add to basket
Replacement screw cap for Bürkle's electrically conductive space saving canisters with DIN50 thread.
  • Made of electrically conductive HDPE
  • Suitable for DIN50 thread
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material HDPE
Sterility Non-sterile
Thread Size (Metric) 30 mm
Neck Finish 30 mm
Disposable Reusable
Product Type Screw Thread Cap
Autoclavable Not Autoclavable