missing translation for 'onlineSavingsMsg'
Learn More

Verenigde Kunststof Bedrijven™ Alveolit Sealring, GORE Membrane, HDPE, Screw Closure for Valve

Catalog Number 10607425
Click to view available options
Color:
Red
This item is not returnable. View return policy
This item is not returnable. View return policy

For Use With: For pressure relief valve

  • HDPE screw cap DIN 51
  • Color - Red
  • Pressure Relief Valve
  • Gore membrane
  • Flood cover
  • DIN 51
Product Type Screw Closure
Description Screwcap DIN 61 Grey
Material HDPE
Color Red
For Use With (Equipment) For pressure relief valve
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'