missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCRIB Recombinant Rabbit Monoclonal Antibody (HL1639)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547116
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.
Specifications
| SCRIB | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q14160 | |
| SCRIB | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SCRIB. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1639 | |
| Unconjugated | |
| SCRIB | |
| AI118201; circletail; Crc; CRIB; CRIB1; hScrib; Kiaa0147; LAP4; mKIAA0147; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; RGD1565055; S Vartul; SCRB1; Scrib; Scrib1; Scribble; scribble homolog 1; scribbled homolog; scribbled homolog (Drosophila); scribbled planar cell polarity protein; Vartul | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23513 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction