missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCRIB Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142476
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_874365.3; NP_056171.3). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.
Specifications
| SCRIB | |
| Polyclonal | |
| Unconjugated | |
| SCRIB | |
| AI118201; circletail; Crc; CRIB; CRIB1; hScrib; Kiaa0147; LAP4; mKIAA0147; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; RGD1565055; S Vartul; SCRB1; Scrib; Scrib1; Scribble; scribble homolog 1; scribbled homolog; scribbled homolog (Drosophila); scribbled planar cell polarity protein; Vartul | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 23513 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q14160 | |
| SCRIB | |
| Peptide with sequence C-PEGPGKEKELPGQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction