missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCRIB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27083-1-AP-150UL
This item is not returnable.
View return policy
Description
SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.Specifications
| SCRIB | |
| Polyclonal | |
| Unconjugated | |
| SCRIB | |
| CRIB1, hScrib, KIAA0147, LAP4, Protein LAP4, Protein scribble homolog, SCRB1, SCRIB, SCRIB1, Scribble, scribbled homolog (Drosophila), Vartul | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105782, 23513 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14160, Q80U72 | |
| Scrib | |
| SCRIB Fusion Protein Ag25795 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction