missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCRN1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17700814
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17700814 200 μL 200µL
17713254 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17700814 Supplier Invitrogen™ Supplier No. PA5119405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SCRN1 Polyclonal Antibody for Western Blot, IHC (P)

This gene likely encodes a member of the secernin family of proteins. A similar protein in rat functions in regulation of exocytosis in mast cells. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SCRN1
Gene Accession No. Q12765, Q9CZC8
Gene Alias 2810019K23Rik; 6330535A03Rik; AI852905; Kiaa0193; mKIAA0193; Scrn1; secernin 1; secernin-1; SES1
Gene Symbols SCRN1
Host Species Rabbit
Immunogen Recombinant protein Secernin 1. The antigen corresponds to amino acid range 2-414 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69938, 9805
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.