missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCUBE1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77185MFV610
This item is not returnable.
View return policy
Description
SCUBE1 Polyclonal antibody specifically detects SCUBE1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| SCUBE1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| signal peptide, CUB and EGF-like domain-containing protein 1, signal peptide, CUB domain, EGF-like 1, signal peptide-CUB domain-EGF-related 1 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the center of human SCUBE1. The immunogen is located within amino acids 460 - 510 of SCUBE1. Amino Acid Squence: CSDAPTTPIKQKAR | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 80274 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction