missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCX Antibody (PCRP-SCXA-1D2) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14175JF635
This item is not returnable.
View return policy
Description
SCX Monoclonal antibody specifically detects SCX in Human samples. It is validated for Flow Cytometry,Immunocytochemistry/ Immunofluorescence
Specifications
| SCX | |
| Monoclonal | |
| Janelia Fluor 635 | |
| BHLHA41, BHLHa48, Class A Basic Helix-Loop-Helix Protein 41, Class A Basic Helix-Loop-Helix Protein 48, Class II BHLH Protein Scleraxis, Scleraxis Basic Helix-Loop-Helix Transcription Factor, Scleraxis BHLH Transcription Factor, Scleraxis Homolog (Mouse), Scleraxis Homolog A, Scleraxis Homolog A (Mouse), Scleraxis Homolog B, Scleraxis Homolog B (Mouse), SCXA, SCXB | |
| Recombinant full-length human SCX protein (Uniprot: Q7RTU7) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| PCRP-SCXA-1D2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 642658 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction