Learn More
SCYL1BP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97159MFV450
Product Code. 18497408
  • € 428.00 / 0.10mL
Description
SCYL1BP1 Polyclonal antibody specifically detects SCYL1BP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SCYL1BP1
Polyclonal
Store at 4C in the dark.
Immunohistochemistry (Paraffin)
mFluor Violet 450 SE