missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCYL1BP1 Antibody (OTI2H6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03929
This item is not returnable.
View return policy
Description
SCYL1BP1 Monoclonal specifically detects SCYL1BP1 in Human samples. It is validated for Western Blot.
Specifications
| SCYL1BP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| GORAB | |
| Human recombinant protein fragment corresponding to amino acids 35-394 of human GORAB (NP_689494) produced in E.coli. | |
| Affinity Purified | |
| RUO | |
| 92344 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| OTI2H6 | |
| Unconjugated | |
| FLJ11752, golgin, RAB6-interacting, GONTKL-binding protein 1, hNTKL-BP1, MGC51263, MGC70512, N-terminal kinase-like-binding protein 1, NTKL-BP1, NTKLBP1SCY1-like 1-binding protein 1, RAB6-interacting golgin, SCY1-like 1 binding protein 1, SCYL1-BP1, SCYL1BP1SCYL1-binding protein 1 | |
| Mouse | |
| 44.8 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction