missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCYL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554317
This item is not returnable.
View return policy
Description
Immunogen sequence: LNVTPTVRPD ADQMTKIPFF DDVGAVTLQY FDTLFQRDNL QKSQFFKGLP KVLPKLPKRV IVQRILPCLT SEFVNPDMVP FVLPNVLLIA EECTKEEYVK LILPELGPVF KQQEPIQILL IFLQKMDLLL TKTPPDEIKN S Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
SCYL2 is a component of AP2-containing clathrin coated structures at the plasma membrane or of endocytic coated vesicles. SCYL2 is a probable serine/threonine-protein kinase that phosphorylates, in vitro, the beta2-subunit of the plasma membrane adapter complex AP2 and other proteins in presence of poly-L-lysine. SCYL2 may also regulate clathrin-dependent trafficking between the TGN and/or the endosomal system.
Specifications
| SCYL2 | |
| Polyclonal | |
| Unconjugated | |
| Scyl2 | |
| AU015688; AW536445; BC030932; Coated vesicle-associated kinase of 104 kDa; CVAK104; D10Ertd802e; KIAA1360; RGD1305665; SCY1 like pseudokinase 2; SCY1-like 2; SCY1-like 2 (S. cerevisiae); SCY1-like protein 2; SCY1-like, kinase-like 2; SCYL2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55681 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P3W7 | |
| Scyl2 | |
| Recombinant protein corresponding to Human SCYL2. Recombinant protein control fragment (Product #RP-91441). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction