Learn More
SCYL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12325-1-AP-20UL
This item is not returnable.
View return policy
Description
MAD2L1 is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. MAD2L1 is related to the MAD2L2 gene located on chromosome 1. A MAD2 pseudogene has been mapped to chromosome 14.Specifications
| SCYL2 | |
| Polyclonal | |
| Unconjugated | |
| SCYL2 | |
| CVAK104, KIAA1360, SCY1 like 2 (S. cerevisiae), SCY1 like protein 2, SCYL2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 213326, 314717, 55681 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6P3W7, Q8CFE4 | |
| Scyl2 | |
| SCYL2 Fusion Protein Ag2988 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |