missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDF2L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571817
This item is not returnable.
View return policy
Description
SDF2L1 Polyclonal Antibody for Western Blot, IHC (P), Flow
The specific function of the protein remains unknown.
Specifications
| SDF2L1 | |
| Polyclonal | |
| Unconjugated | |
| SDF2L1 | |
| dihydropyrimidinase-like 2; OTTHUMT00000075032; PWP1-interacting protein 8; Sdf2l1; SDF2-like 1; SDF2-like protein 1; stromal cell derived factor 2 like 1; stromal cell-derived factor 2-like 1; stromal cell-derived factor 2-like protein 1; UNQ1941/PRO4424 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 23753, 64136 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9ESP1, Q9HCN8 | |
| SDF2L1 | |
| KLH conjugated synthetic peptide between 160-189 amino acids from the C-terminal region of human SDF2L1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction