missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SDF2L1 Polyclonal Antibody for IHC
Specifications
Specifications
| Antigen | SDF2L1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SDF2L1 |
| Gene Accession No. | Q9HCN8 |
| Gene Alias | dihydropyrimidinase-like 2; OTTHUMT00000075032; PWP1-interacting protein 8; Sdf2l1; SDF2-like 1; SDF2-like protein 1; stromal cell derived factor 2 like 1; stromal cell-derived factor 2-like 1; stromal cell-derived factor 2-like protein 1; UNQ1941/PRO4424 |
| Gene Symbols | SDF2L1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
