missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDF2L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20888-1-AP-150UL
This item is not returnable.
View return policy
Description
The specific function of the protein remains unknown.Specifications
| SDF2L1 | |
| Polyclonal | |
| Unconjugated | |
| SDF2L1 | |
| PWP1 interacting protein 8, SDF2 like protein 1, SDF2L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23753, 64136, 680945 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ESP1, Q9HCN8 | |
| Sdf2l1 | |
| SDF2L1 Fusion Protein Ag14160 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction