missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598828
This item is not returnable.
View return policy
Description
SDF4 Polyclonal Antibody for Western Blot, ELISA
This gene encodes a stromal cell derived factor that is a member of the CREC protein family. The encoded protein contains six EF-hand motifs and calcium-binding motifs. This protein localizes to the Golgi lumen and may be involved in regulating calcium dependent cellular activities.
Specifications
| SDF4 | |
| Polyclonal | |
| Unconjugated | |
| SDF4 | |
| 45 kDa calcium-binding protein; cab45; PSEC0034; Sdf4; SDF-4; stromal cell derived factor 4; stromal cell-derived factor 4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 51150 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9BRK5 | |
| SDF4 | |
| Recombinant Human 45 kDa calcium-binding protein (37-362AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction