missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDH Assembly Factor 1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41176MFV610
This item is not returnable.
View return policy
Description
SDH Assembly Factor 1 Polyclonal antibody specifically detects SDH Assembly Factor 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| SDH Assembly Factor 1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| LYR Motif Containing 8, LYR Motif-Containing Protein 8, LYRM8, SDHAF1, Succinate Dehydrogenase Assembly Factor 1, Mitochondrial, Succinate Dehydrogenase Complex Assembly Factor 1 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SDHAF1. The immunogen is located within the last 50 amino acids of SDHAF1. Amino Acid Squence: DSPRNPHDSTGAPETRPD | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 644096 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction