missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHAF1 Monoclonal Antibody (OTI1E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809691S
This item is not returnable.
View return policy
Description
SDHAF1 Monoclonal antibody specifically detects SDHAF1 in Human samples. It is validated for Western BlotSpecifications
| SDHAF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BY41 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55869 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1E6 | |
| Unconjugated | |
| HDAC8 | |
| 0610010E21Rik; AI430885; AW490662; LYR motif containing 8; LYR motif-containing protein 8; LYRM8; SDH assembly factor 1; Sdhaf1; Succinate dehydrogenase assembly factor 1, mitochondrial; succinate dehydrogenase complex assembly factor 1 | |
| Human recombinant protein fragment corresponding to amino acids 1-243 of human HDAC8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction