missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDHAF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525308
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
SDHAF1 (Succinate dehydrogenase complex assembly factor 1) is one of the subunits of the succinate dehydrogenase (SDH) complex (or complex II) of the mitochondrial respiratory chain. SDHAF1 resides in the mitochondria, and is essential for SDH assembly, but does not physically associate with the complex in vivo. Mutations in this gene are associated with SDH-defective infantile leukoencephalopathy (mitochondrial complex II deficiency). Unlike the related protein SDHAF2, SDHAF1 is not thought to be a tumor suppressor.
Specifications
| SDHAF1 | |
| Polyclonal | |
| Unconjugated | |
| SDHAF1 | |
| 0610010E21Rik; AI430885; AW490662; LYR motif containing 8; LYR motif-containing protein 8; LYRM8; SDH assembly factor 1; Sdhaf1; Succinate dehydrogenase assembly factor 1, mitochondrial; succinate dehydrogenase complex assembly factor 1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 644096 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A6NFY7 | |
| SDHAF1 | |
| KLH conjugated synthetic peptide between 34-62 amino acids from the central region of human SDHAF1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction