missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHB Antibody (SDHB/2382), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08537AF700
This item is not returnable.
View return policy
Description
SDHB Monoclonal antibody specifically detects SDHB in Human samples. It is validated for Western Blot, Protein Array
Specifications
| SDHB | |
| Monoclonal | |
| Alexa Fluor 700 | |
| EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip) | |
| Recombinant fragment (around aa 165-273) of human SDHB protein (exact sequence is proprietary) (Uniprot: P21912) | |
| 100 μg | |
| Cancer | |
| 6390 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Protein Array | |
| SDHB/2382 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction