Learn More
SDMA, MedChemExpress
Product Code. 16493348
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
Product Code. 16493348
Brand: Medchem Express HY101410
MedChemExpress SDMA (Symmetric dimethylarginine) is an endogenous inhibitor of nitric oxide (NO) synthase activity.
SDMA (Symmetric dimethylarginine) is an endogenous inhibitor of nitric oxide (NO) synthase activity.
Specifications
Chemical Name or Material SDMA
Physical Form Powder
CAS 30344-00-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C8H18N4O2
For Use With (Application) COVID-19-immunoregulation
Synonym Symmetric dimethylarginine NG,NG'-Dimethyl-L-arginine
Solubility Information DMSO : 67.5 mg/mL (333.75 mM; Need ultrasonic) ∣H2O : 50 mg/mL (247.22 mM; ultrasonic and warming and heat to 60°C)
SMILES N[C@@H](CCC/N=C(NC)/NC)C(O)=O
Molecular Weight (g/mol) 202.25
Formula Weight 202.25
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only