missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDR9C7 Antibody (OTI4B5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74036F
This item is not returnable.
View return policy
Description
SDR9C7 Monoclonal specifically detects SDR9C7 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| SDR9C7 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SDR9C7 | |
| Full length recombinant protein of human SDR9C7 (NP_683695) produced in E.coli. | |
| 0.1 mL | |
| 121214.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry | |
| OTI4B5 | |
| Flow Cytometry, Immunohistochemistry | |
| EC 1.1.1, EC 1.1.1.-, FLJ16333, MGC126600, Orphan short-chain dehydrogenase/reductase, RDH-S, RDHSMGC126602, SDR-Oorphan short-chain dehydrogenase/ reductase, SDROretinol dehydrogenase similar protein, short chain dehydrogenase/reductase family 9C, member 7, short-chain dehydrogenase/reductase family 9C member 7 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction