missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF39 Monoclonal Antibody (OTI5F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505915
This item is not returnable.
View return policy
Description
This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| RNF39 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H2S5 | |
| RNF39 | |
| Full length human recombit protein of human RNF39 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5F5 | |
| Unconjugated | |
| RNF39 | |
| HZF, HZFW, LIRF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 386454, 80352 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction