Learn More
SDS Mouse anti-Human, Clone: OTI1C8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503994
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SDS encodes one of three enzymes that are involved in metabolizing serine and glycine. L-serine dehydratase converts L-serine to pyruvate and ammonia and requires pyridoxal phosphate as a cofactor. The encoded protein can also metabolize threonine to NH4+ and 2-ketobutyrate. The encoded protein is found predomitly in the liver.Specifications
| SDS | |
| Monoclonal | |
| Unconjugated | |
| SDS | |
| SDH | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10993 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1C8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P20132 | |
| SDS | |
| Full length human recombit protein of human SDS produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.