missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDS3 Antibody, DyLight 650, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-257C
This item is not returnable.
View return policy
Description
SDS3 Polyclonal antibody specifically detects SDS3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SDS3 | |
| Polyclonal | |
| Western Blot | |
| 45 kDa Sin3-associated polypeptide, FLJ00052, SAP45suppressor of defective silencing 3 homolog (SDS3, S. cerevisiae), SDS3MGC104711, sin3 histone deacetylase corepressor complex component SDS3, sin3A-associated protein, 45kDa, suppressor of defective silencing 3 homolog (S. cerevisiae), Suppressor of defective silencing 3 protein homolog | |
| The immunogen recognized by this antibody maps to a region between residues 175 and 225 of human Suppressor of Defective Silencing 3 using the numbering given in entry NP_071936.2 (GeneID 64426). | |
| 0.1 mL | |
| DNA Repair | |
| 64426 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction