missing translation for 'onlineSavingsMsg'
Learn More

SDSL Antibody (OTI1A8), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-45978

Product Code. 18159258

  • € 577.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry

SDSL Monoclonal specifically detects SDSL in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SDSL
Monoclonal
1 mg/mL
Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:150Flow 1:100
EC 4.3.1.17, EC 4.3.1.19, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDH, SDH 2, SDS-RS1, Serine dehydratase 2, serine dehydratase related sequence 1, serine dehydratase-like, TDH
Mouse
34.5 kDa
0.1 mL
metabolism
113675
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry
OTI1A8
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
SDSL
Full length human recombinant protein of human SDSL(NP_612441) produced in HEK293T cell.
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only