missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDSL Antibody (OTI1A8), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74045PECY7
This item is not returnable.
View return policy
Description
SDSL Monoclonal specifically detects SDSL in Human samples. It is validated for Flow Cytometry.
Specifications
| SDSL | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| SDSL | |
| Full length recombinant protein of human SDSL (NP_612441) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI1A8 | |
| Flow Cytometry | |
| EC 4.3.1.17, EC 4.3.1.19, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDH, SDH 2, SDS-RS1, Serine dehydratase 2, serine dehydratase related sequence 1, serine dehydratase-like, TDH | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 113675.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction