missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDSL Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542766
This item is not returnable.
View return policy
Description
Peptide sequence: ALAAIYSGLL RRLQAEGCLP PSLTSVVVIV CGGNNINSRE LQALKTHLGQ Sequence homology: Horse: 79%; Human: 100%; Rabbit: 86%.
SDSL has low serine dehydratase and threonine dehydratase activity.
Specifications
| SDSL | |
| Polyclonal | |
| Unconjugated | |
| SDSL | |
| 4432411H13Rik; AI504310; cSDH; L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH; SDH 2; SDH1; Sds; SDSL; SDS-RS1; Serine dehydratase 2; serine dehydratase like; serine dehydratase related sequence 1; serine dehydratase-like; TDH | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113675 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96GA7 | |
| SDSL | |
| Synthetic peptide directed towards the middle region of human SDSL. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction