missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDSL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16242-1-AP-20UL
This item is not returnable.
View return policy
Description
SDSL has low serine dehydratase and threonine dehydratase activity.Specifications
| SDSL | |
| Polyclonal | |
| Unconjugated | |
| SDSL | |
| L serine deaminase, L threonine dehydratase, SDH 2, SDS RS1, SDSL, Serine dehydratase 2, serine dehydratase like, TDH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 113675, 257635 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R238, Q96GA7 | |
| SDSL | |
| SDSL Fusion Protein Ag9114 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction