Learn More
SeaKem™ GTG™ Agarose, Lonza™
Product Code. 11560727
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11560727 1 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11560727 Supplier Lonza Supplier No. 50075
Standard gelling temperature. Lonza SeaKem™ GTG™ Agarose is specifically designed for preparative DNA electrophoresis and tested for presence of proteases, ligases and nucleases.
  • High gel strength agarose that resolves DNA fragments greater than 1,000 bp
  • DNA recovered from agarose gels are reliably digested and ligated
  • Performance tested to ensure complete compatibility with routine molecular biology techniques
  • Certificate of performance containing end-use test information is provided
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kg