Learn More
asecos™ Sealant Mat
Product Code. 17896356 Shop All asecos Products
Change view
Click to view available options
Width (Metric):
400.00 mm
600.00 mm
800.00 mm
1000.00 mm
1200.00 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
17896356 400.00 mm Each
17806536 600.00 mm Each
17886406 800.00 mm Each
17866466 1000.00 mm Each
17826536 1200.00 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17896356 Supplier asecos™ Supplier No. 9902
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
The Sealant Mat provides rapid protection in case of leaking liquids.
  • Prevents in case of fire or emergency a contamination of the surroundings with waterpollutant liquids
  • Incl. protection film with integral handles
  • Resistant to water, hydrocarbons and most chemicals
  • The extremely dense, flexible material adhering to the underside adapts perfectly to the nature of the ground
  • Can be used several times
Specifications
Color Yellow
Material Polyurethane
Type Barrier Spill Matting
Length (English) 15.75 in.
Length (Metric) 400.00 mm
Width (English) 15.75 in.
Width (Metric) 400.00 mm
Height (English) 0.31 in.
Height (Metric) 8.00 mm
Weight (English) 3.86 lb.
Weight (Metric) 1.75 kg