missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One SealPlate™ Microplate Seal
Click to view available options
For Use With (Equipment):
Standard Microwell Plates
Unit Size:
Pack of 200
Description
- Adhesive film designed to cover microplates
- Protect samples from evaporation and contamination
- Non-sterile
- Convenient and easy to apply
Specifications
Specifications
| Color | Transparent |
| Sterility | Non-sterile |
| No. per Pack | 100 |
| Dimensions (L x W) | 120 x 80 mm |
| For Use With (Equipment) | Standard Microwell Plates |
| Plate Compatibility | Standard Microwell Plate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction