Learn More
Lenz Laborglasintrumente Seals for Ball Flanges
Product Code. p-7222544
Click to view available options
Includes:
Lab Flange 40
Lab Flange 50
Unit Size:
Each
Product Code. 11923331
Brand: Lenz Laborglasintrumente 06248840
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Seals for ball flanges KF 15-KF 50
  • Material: PTFE
Specifications
Includes Lab Flange 40
Material PTFE
Quantity 1 Ea.