missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Hemagglutinin Antibody (7H12F6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41328H
This item is not returnable.
View return policy
Description
Seasonal H1N1 Hemagglutinin Monoclonal antibody specifically detects Seasonal H1N1 Hemagglutinin in Influenza Virus N1, Virus samples. It is validated for Western Blot, ELISA
Specifications
| Seasonal H1N1 Hemagglutinin | |
| Monoclonal | |
| HRP | |
| Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
| Antibody was raised against a synthetic peptide from the Hemagglutinin protein. Amino Acid Squence: PNIGDQKALTHTEN | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Influenza virus N1, Virus | |
| Purified |
| Western Blot, ELISA | |
| 7H12F6 | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction