missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Hemagglutinin Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41108AF700
This item is not returnable.
View return policy
Description
Seasonal H1N1 Hemagglutinin Polyclonal antibody specifically detects Seasonal H1N1 Hemagglutinin in Influenza Virus N1, Virus samples. It is validated for ELISA
Specifications
| Seasonal H1N1 Hemagglutinin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 700 | |
| Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
| Antibody was raised against a synthetic peptide from the Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41104, and NBP2-41106. This antibody is a cognate pair with product number NBP2-41109. The immunogen is located within amino acids 150 - 200 of Seasonal H1N1 Hemagglutinin. Amino Acid Squence: TGKNGLYPN | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Influenza virus N1, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction