missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptopodin Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-39100AF594
This item is not returnable.
View return policy
Description
Synaptopodin Polyclonal specifically detects Synaptopodin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Synaptopodin | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| KIAA1029, synaptopodin | |
| Rabbit | |
| Peptide affinity purified | |
| Primary | |
| At least three isoforms of SYNPO are known to exist; this antibody will detect all three isoforms. SYNPO antibody is predicted to not cross-react with other SYNPO family member proteins. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SYNPO | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SYNPO. The immunogen is located within the last 50 amino acids of SYNPO. Amino Acid Squence: SPSRAWSPRAKQAPRPS | |
| 0.1 mL | |
| 11346 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction