missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Hemagglutinin Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41104G
This item is not returnable.
View return policy
Description
Seasonal H1N1 Hemagglutinin Polyclonal antibody specifically detects Seasonal H1N1 Hemagglutinin in Influenza virusN1, Virus samples. It is validated for Western Blot, ELISA
Specifications
| Seasonal H1N1 Hemagglutinin | |
| Polyclonal | |
| Western Blot, ELISA | |
| Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
| Antibody was raised against a synthetic peptide from the Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41106 and NBP2-41108. This antibody is a cognate pair with product NBP2-41105. The immunogen is located within amino acids 120 - 170 of Seasonal H1N1 Hemagglutinin. Amino Acid Squence: PNHTVTGVSAS | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Influenza virus N1, Virus, Rat | |
| Purified |
| Western Blot, ELISA | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction