missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Hemagglutinin Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41106MFV610
This item is not returnable.
View return policy
Description
Seasonal H1N1 Hemagglutinin Polyclonal antibody specifically detects Seasonal H1N1 Hemagglutinin in Influenza virusN1, Virus samples. It is validated for ELISA
Specifications
| Seasonal H1N1 Hemagglutinin | |
| Polyclonal | |
| ELISA | |
| Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
| Antibody was raised against a synthetic peptide from the Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41104 and NBP2-41108. This antibody is a cognate pair with antibody NBP2-41107. The immunogen is located within amino acids 180 - 230 of Seasonal H1N1 Hemagglutinin. Amino Acid Squence: PNIGDQKALTHTEN | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Influenza virus N1, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction