Learn More
Seasonal H1N1 Neuraminidase Antibody (10C5E11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-41330
 View more versions of this product
Product Code. 18105939
  • € 509.00 / 0.10mg
Description
Seasonal H1N1 Neuraminidase Monoclonal specifically detects Seasonal H1N1 Neuraminidase in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Seasonal H1N1 Neuraminidase
Monoclonal
1 mg/mL
ELISA 1 μg/mL
EC=3.2.1.18, Neuraminidase
Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein.
0.1 mg
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
ELISA
10C5E11
Unconjugated
PBS with 0.02% Sodium Azide
Mouse
Affinity Purified
RUO
Influenza virus, Virus
IgG2b