missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Neuraminidase Antibody (10C5F12), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41331MFV610
This item is not returnable.
View return policy
Description
Seasonal H1N1 Neuraminidase Monoclonal antibody specifically detects Seasonal H1N1 Neuraminidase in Influenza virusN1, Virus samples. It is validated for ELISA
Specifications
| Seasonal H1N1 Neuraminidase | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Influenza virus N1, Virus | |
| Purified |
| ELISA | |
| 10C5F12 | |
| ELISA | |
| EC=3.2.1.18, Neuraminidase | |
| Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction