missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 NS1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41113AF647
This item is not returnable.
View return policy
Description
Seasonal H1N1 NS1 Polyclonal antibody specifically detects Seasonal H1N1 NS1 in Influenza Virus N1 samples. It is validated for ELISA
Specifications
| Seasonal H1N1 NS1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Antibody was raised against a synthetic peptide from the seasonal Influenza NS1 protein. The immunogen is located within the last 50 amino acids of Seasonal H1N1 Nonstructural Protein 1. Amino Acid Squence: SSNETGGPPFTTT | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Influenza virus N1 | |
| Purified |
| ELISA | |
| Alexa Fluor 647 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction