missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Nucleocapsid Protein Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41114AF488
This item is not returnable.
View return policy
Description
Seasonal H1N1 Nucleocapsid Protein Polyclonal antibody specifically detects Seasonal H1N1 Nucleocapsid Protein in Influenza Virus N1, Virus samples. It is validated for ELISA
Specifications
| Seasonal H1N1 Nucleocapsid Protein | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 488 | |
| Nucleocapsid protein, Nucleoprotein, Protein N | |
| Antibody was raised against a synthetic peptide from the seasonal H1N1 NP protein. The immunogen is located within amino acids 350 - 400 of Seasonal H1N1 Nucleocapsid Protein. Amino Acid Squence: NMDNMGSS | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Influenza virus N1, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction