missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC11A Mouse anti-Human, Clone: OTI1A12, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811462
This item is not returnable.
View return policy
Description
Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.Specifications
| SEC11A | |
| Monoclonal | |
| Unconjugated | |
| SEC11A | |
| 1810012E07Rik, SEC11L1, SPC18, SPCS4A, sid2895 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23478 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1A12 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P67812 | |
| SEC11A | |
| Full length human recombit protein of human SEC11A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction